Thermo Scientific Chemicals

Pyrogallol, 98+%, Thermo Scientific Chemicals

Catalog number: A13405.22
100 g, Each
Thermo Scientific Chemicals

Pyrogallol, 98+%, Thermo Scientific Chemicals

Catalog number: A13405.22
100 g, Each
Quantity
Catalog number: A13405.22
Price (USD)
Price: 64.50
Online price: 54.65
Your price:
Quantity
-

Chemical Identifiers

CAS
87-66-1
IUPAC Name
benzene-1,2,3-triol
Molecular Formula
C6H6O3
InChI Key
WQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILES
OC1=CC=CC(O)=C1O
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥98.0%
Melting Point (clear melt)
131-136?C

Description

Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal-complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal-complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

Solubility
Soluble in ethanol, water and ether. Slightly soluble in chloroform.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, alkalis, metal oxides, ammonia, iodine, lime water, potassium permanganate and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text