L-Aspartic acid, 98+%
L-Aspartic acid, 98+%
L-Aspartic acid, 98+%
Thermo Scientific Chemicals

L-Aspartic acid, 98+%

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol
数量:
250 g
1000 g
5000 g
製品番号(カタログ番号) A13520.0B
または、製品番号A13520-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS56-84-8
IUPAC Name2-aminobutanedioic acid
Molecular FormulaC4H7NO4
InChI KeyCKLJMWTZIZZHCS-UHFFFAOYNA-N
SMILESNC(CC(O)=O)C(O)=O
さらに表示
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0% (dry wt. basis)
Loss on Drying≤0.5% (105°C)
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
さらに表示
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.

Solubility
Soluble in water(4.5g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Flaig, R.; Koritsanszky, T.; Zobel, D.; Luger, P. Topological Analysis of the Experimental Electron Densities of Amino Acids. 1. d,l-Aspartic Acid at 20 K. J. Am. Chem. Soc. 1998, 120, (10), 2227-2238.
  2. Xiao-Lin Wang.; Ai-Ling Ying.; Wei-Ning Wang. Nanofiltration of l-phenylalanine and l-aspartic acid aqueous solutions. Journal of Membrane Science. 2002, 196, (1), 59-67.
  3. Readily forms ß-lactam derivatives whichare valuable intermediates for the synthesis of a variety of products, including thienamycin: J. Am. Chem. Soc., 102, 6163 (1980); Tetrahedron Lett., 23, 2293 (1982).