Thermo Scientific Chemicals

Benzylideneacetone, 98+%, Thermo Scientific Chemicals

Catalog number: A13537.0B
1000 g, Each
Thermo Scientific Chemicals

Benzylideneacetone, 98+%, Thermo Scientific Chemicals

Catalog number: A13537.0B
1000 g, Each
Quantity
Catalog number: A13537.0B
also known as A13537-0B
Price (USD)
Price: 97.30
Online price: 83.65
Your price:
Quantity
-

Chemical Identifiers

CAS
122-57-6
Form
Crystals or powder or crystalline powder or fused solid or crystalline chunks or clear liquid as melt
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow to yellow
Assay (GC)
≥98.0%

Description

Benzylideneacetone is used chiefly as a scent in the manufacture of perfume, drugs & electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylideneacetone is used chiefly as a scent in the manufacture of perfume, drugs & electroplating.

Solubility
Soluble in alcohol, chloroform, diethyl ether.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text