Benzylideneacetone, 98+%
Benzylideneacetone, 98+%
Benzylideneacetone, 98+%
Benzylideneacetone, 98+%
Thermo Scientific Chemicals

Benzylideneacetone, 98+%

CAS: 122-57-6 | C10H10O | 146.19 g/mol
製品番号(カタログ番号) A13537.30
または、製品番号A13537-30
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS122-57-6
FormCrystals or powder or crystalline powder or fused solid or crystalline chunks or clear liquid as melt
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to yellow
Assay (GC)≥98.0%

Benzylideneacetone is used chiefly as a scent in the manufacture of perfume, drugs & electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylideneacetone is used chiefly as a scent in the manufacture of perfume, drugs & electroplating.

Solubility
Soluble in alcohol, chloroform, diethyl ether.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Miguel A. Esteruelas.; Luis A. Oro.; Cristina Valero. Hydrogenation of benzylideneacetone catalyzed by OsHCl(CO)(PR3)2 (PR3 = P-iso-Pr3, PMe-tert-Bu2): new roles of dihydrogen complexes in homogeneous catalytic hydrogenation. Organometallics. 1992, 11, (10), 3362-3369.
  2. Lubin Luo.; Steven P. Nolan. Enthalpies of reaction of (benzylideneacetone)iron tricarbonyl, (BDA)Fe(CO)3, with phosphine ligands. Thermodynamic insights into iron chemistry. Organometallics. 1992, 11, (10), 3483-3486.