2-(Methoxycarbonyl)benzenesulfonamide, 98%
2-(Methoxycarbonyl)benzenesulfonamide, 98%
2-(Methoxycarbonyl)benzenesulfonamide, 98%
Thermo Scientific Chemicals

2-(Methoxycarbonyl)benzenesulfonamide, 98%

CAS: 57683-71-3 | C8H9NO4S | 215.22 g/mol
製品番号(カタログ番号) A13553.14
または、製品番号A13553-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS57683-71-3
IUPAC Namemethyl 2-sulfamoylbenzoate
Molecular FormulaC8H9NO4S
InChI KeyVSOOBQALJVLTBH-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1S(N)(=O)=O
さらに表示
Identification (FTIR)Conforms
Melting Point123.0-129.0°C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only