Thermo Scientific Chemicals

Dicyclopentadiene, 90+%, stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A13643.AP
500 mL, Each
Thermo Scientific Chemicals

Dicyclopentadiene, 90+%, stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A13643.AP
500 mL, Each
Quantity
Catalog number: A13643.AP
also known as A13643-AP
Price (USD)
Quantity
-

Chemical Identifiers

CAS
77-73-6
IUPAC Name
tricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular Formula
C10H12
InChI Key
HECLRDQVFMWTQS-UHFFFAOYNA-N
SMILES
C1C=CC2C3CC(C=C3)C12
Appearance (Color)
Clear, colourless to pale yellow
Refractive Index
1.5065 - 1.5145 @20?C
Form
liquid
Assay (GC)
> 90.0% (major isomer)

Description

Dicyclopentadiene is used in resins, particularly unsaturated polyester resins. It plays a major role in inks, adhesives and paints. It is also used as a monomer in polymerization reactions. It is a precursor for the preparation of endo-tetrahydrodicyclopentadiene, which reacts with aluminum chloride at higher temperature to give adamantine. It undergoes a retro-Diels-Alder reaction to yield cyclopentadiene, which acts as ligand in inorganic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dicyclopentadiene is used in resins, particularly unsaturated polyester resins. It plays a major role in inks, adhesives and paints. It is also used as a monomer in polymerization reactions. It is a precursor for the preparation of endo-tetrahydrodicyclopentadiene, which reacts with aluminum chloride at higher temperature to give adamantine. It undergoes a retro-Diels-Alder reaction to yield cyclopentadiene, which acts as ligand in inorganic chemistry.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text