3-(4-Methoxyphenyl)propionic acid, 98%
3-(4-Methoxyphenyl)propionic acid, 98%
3-(4-Methoxyphenyl)propionic acid, 98%
3-(4-Methoxyphenyl)propionic acid, 98%
Thermo Scientific Chemicals

3-(4-Methoxyphenyl)propionic acid, 98%

CAS: 1929-29-9 | C10H12O3 | 180.203 g/mol
製品番号(カタログ番号) A13655.14
または、製品番号A13655-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1929-29-9
IUPAC Name3-(4-methoxyphenyl)propanoic acid
Molecular FormulaC10H12O3
InChI KeyFIUFLISGGHNPSM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CCC(O)=O)C=C1
さらに表示
Appearance (Color)White to cream to brown
FormCrystalline powder or powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (UK-sourced material)
Assay (Silylated GC)≥97.5% (UK-sourced material)
Assay from Supplier's CofA≥97.5% (US-sourced material)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only