2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
2,4-Difluoroaniline, 99%
Thermo Scientific Chemicals

2,4-Difluoroaniline, 99%

CAS: 367-25-9 | C6H5F2N | 129.11 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13673.36
または、製品番号A13673-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS367-25-9
IUPAC Name2,4-difluoroaniline
Molecular FormulaC6H5F2N
InChI KeyCEPCPXLLFXPZGW-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1F
さらに表示
Appearance (Color)Clear colorless to pale yellow to pale brown or pink or purple
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5060-1.5090 @ 20°C
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Karlyn A. Schnapp.; Russell Poe.; Elisa Leyva.; N. Soundararajan, Matthew S. Platz. Exploratory photochemistry of fluorinated aryl azides. Implications for the design of photoaffinity labeling reagents. Bioconjugate Chem. 1993, 4 (2), 172-177.
  2. Yaofeng Chen.; Changtao Qian.; Jie Sun. Fluoro-Substituted 2,6-Bis(imino)pyridyl Iron and Cobalt Complexes: High-Activity Ethylene Oligomerization Catalysts. Organometallics. 2003, 22 (6), 1231-1236.