1-Chloro-4-fluorobenzene, 98%
1-Chloro-4-fluorobenzene, 98%
1-Chloro-4-fluorobenzene, 98%
1-Chloro-4-fluorobenzene, 98%
Thermo Scientific Chemicals

1-Chloro-4-fluorobenzene, 98%

CAS: 352-33-0 | C6H4ClF | 130.55 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A13690.36
または、製品番号A13690-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS352-33-0
IUPAC Name1-chloro-4-fluorobenzene
Molecular FormulaC6H4ClF
InChI KeyRJCGZNCCVKIBHO-UHFFFAOYSA-N
SMILESFC1=CC=C(Cl)C=C1
さらに表示
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4930-1.4970 @ 20?C
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4'-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 '-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4′-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 ′-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Lithiation with LDA occurs mainly ortho to F; subsequent carboxylation gives 5-chloro-2-fluorobenzoic acid in moderate yield: Tetrahedron Lett., 33, 7495 (1992). The selectivity is improved by the use of the superbasic combination of n-BuLi and KO-t-Bu, and the yield is increased to 86%: Tetrahedron Lett., 37, 6551 (1996).