1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%
1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%
1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%
Thermo Scientific Chemicals

1,3-Dimethylbarbituric acid, 99% (dry wt.), water <6%

CAS: 769-42-6 | C6H8N2O3 | 156.141 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A13692.0B
または、製品番号A13692-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS769-42-6
IUPAC Name1,3-dimethyl-1,3-diazinane-2,4,6-trione
Molecular FormulaC6H8N2O3
InChI KeyVVSASNKOFCZVES-UHFFFAOYSA-N
SMILESCN1C(=O)CC(=O)N(C)C1=O
さらに表示
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (dry wt. basis)
Identification (FTIR)Conforms
Assay (Silylated GC)≥98.5%
さらに表示
1,3-Dimethylbarbituric acid is used as a catalyst in the Knoevenagel condensation of a series of aromatic aldehydes. It is also used in the synthesis of 5-aryl-6-(alkyl- or aryl-amino)-1,3-dimethylfuro [2,3-d]pyrimidine derivatives and enantioselective synthesis of isochromene pyrimidinedione derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only