Thermo Scientific Chemicals

Ethyl trifluoroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A13717.18
50 g, Each
Thermo Scientific Chemicals

Ethyl trifluoroacetoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A13717.18
50 g, Each
Quantity
Catalog number: A13717.18
also known as A13717-18
Price (USD)
Price: 149.00
Online price: 128.65
Your price:
Quantity
-

Chemical Identifiers

CAS
372-31-6
IUPAC Name
ethyl 4,4,4-trifluoro-3-oxobutanoate
Molecular Formula
C6H7F3O3
InChI Key
OCJKUQIPRNZDTK-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C(F)(F)F
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.3750-1.3790 @ 20?C

Description

Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trifluoroacetoacetate is used as an intermediate in synthetic chemistry. It is also used as an intermediate for agro chemicals and pharmaceuticals. Further, it is used in the preparation of fluorinated 2-thiouracil analogs as antithyroid agents.

Solubility
Miscible with water, ethanol, benzene and organic solvents.

Notes
Incompatible with oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text