1-Butylboronic acid, 98%
1-Butylboronic acid, 98%
1-Butylboronic acid, 98%
Thermo Scientific Chemicals

1-Butylboronic acid, 98%

CAS: 4426-47-5 | C4H11BO2 | 101.94 g/mol
製品番号(カタログ番号) A13725.03
または、製品番号A13725-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4426-47-5
IUPAC Namebutylboronic acid
Molecular FormulaC4H11BO2
InChI KeyQPKFVRWIISEVCW-UHFFFAOYSA-N
SMILESCCCCB(O)O
さらに表示
Proton NMRConforms to structure
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5%
FormCrystals or powder or crystalline powder or flakes
Identification (FTIR)Conforms
1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butylboronic acid is used as a reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry and to prepare chiral oxazaborolidines. It is a precursor to unsymmetric borinic acids, which are inhibitors of serine proteases. Can be used as a transport carrier in membrane-based sugar separations and as an analytical reagent in the determination of serum glucose.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is hygroscopic in nature. Incompatible with oxidizing agents. Store under dry inert gas.
RUO – Research Use Only

General References:

  1. Franck Picard.; Tobias Schulz.; Rolf W. Hartmann. 5-Phenyl substituted 1-methyl-2-pyridones and 4'-substituted biphenyl-4-carboxylic acids. synthesis and evaluation as inhibitors of steroid-5α-reductase type 1 and 2. Bioorganic & Medicinal Chemistry. 2002, 10 (2), 437-448.
  2. Zhao Yongliang.; Zhao Fengying.; Li Qiang.; Gao Deqing. Synthesis, Characterization and Fluorescence Properties of Europium, Terbium Complexes with Biphenyl-4-Carboxylic Acid and o-Phenanthroline. Journal of Rare Earths. 2006, 24 (1), 18-22.
  3. Reagent for preparation of volatile derivatives of diols, e.g. carbohydrates, for GC and mass spectrometry: J. Chromat., 54, 193 (1971); J. Chromat. Sci., 9, 18 (1971); Gas Chromat., 129 (1968).
  4. In combination with (S)-(-)-ɑ,ɑ-Diphenyl prolinol, L09217, or its enantiomer (9218), oxazaborolidine catalysts are generated in situ, for use in highly enantioselective reductions: Tetrahedron Lett., 33, 4141 (1992). The same system has been employed in the synthesis of chiral monosubstituted oxiranes: Tetrahedron Lett., 34, 5227 (1993). See also (S)-2-Methyl-CBS-oxazaborolidine monohydrate, L09219, and Appendix 5.