Acetone-1,3-dicarboxylic acid, 97%
Acetone-1,3-dicarboxylic acid, 97%
Acetone-1,3-dicarboxylic acid, 97%
Thermo Scientific Chemicals

Acetone-1,3-dicarboxylic acid, 97%

CAS: 542-05-2 | C5H6O5 | 146.098 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A13742.0B
または、製品番号A13742-0B
価格(JPY)
-
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS542-05-2
IUPAC Name3-oxopentanedioate
Molecular FormulaC5H4O5
InChI KeyOXTNCQMOKLOUAM-UHFFFAOYSA-L
SMILES[O-]C(=O)CC(=O)CC([O-])=O
さらに表示
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0%
Identification (FTIR)Conforms
Loss on Drying≤3.0% (50°C/constant weight)
Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetone-1,3-dicarboxylic acid is used as a building block in organic chemistry. It is also used in the synthesis of hetrocyclic rings and in the Weiss-Cook reaction, which involves the preparation of cis-bicyclo[3.3.0]octane-3,7-dione by reacting with diacyl (1,2 ketone). Its presence in human urine is used as a diagnostic test for the overgrowth of harmful gut flora such as Candida albicans.

Solubility
Soluble in water and alcohol. Slightly soluble in ethyl acetate and ether. Insoluble in chloroform.

Notes
Hygroscopic. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. For use in Robinson's classical one-step synthesis of pseudopelletierine, see: Org. Synth. Coll., 4, 816 (1963):
  2. For a more recent analogous example of the synthesis of azabicyclo[3.3.1]nonan-3-one derivatives, see: J. Med. Chem., 36, 3707 (1993).
  3. Li, C.; Breit, B. Rhodium-Catalyzed Chemo- and Regioselective Decarboxylative Addition of beta-Ketoacids to Allenes: Efficient Construction of Tertiary and Quaternary Carbon Centers. J. Am. Chem. Soc. 2014, 136 (3), 862-865.
  4. Sousa, A.; Araújo, P.; Mateus, N.; de Freitas, V. Deoxyvitisins: a new set of pyrano-3-deoxyanthocyanidins. Tetrahedron Lett. 2013, 54 (35), 4785-4788.