2-Chloro-5-nitrobenzoic acid, 98+%
2-Chloro-5-nitrobenzoic acid, 98+%
2-Chloro-5-nitrobenzoic acid, 98+%
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzoic acid, 98+%

CAS: 2516-96-3 | C7H4ClNO4 | 201.562 g/mol
製品番号(カタログ番号) A13754.36
または、製品番号A13754-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2516-96-3
IUPAC Name2-chloro-5-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyQUEKGYQTRJVEQC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC=C1Cl)[N+]([O-])=O
さらに表示
Appearance (Color)White to pale cream to pale yellow
Melting Point (clear melt)162.0-168.0?C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only