m-Toluic acid, 99%
m-Toluic acid, 99%
m-Toluic acid, 99%
Thermo Scientific Chemicals

m-Toluic acid, 99%

CAS: 99-04-7 | C8H8O2 | 136.15 g/mol
数量:
250 g
1000 g
5000 g
製品番号(カタログ番号) A13785.0I
または、製品番号A13785-0I
価格(JPY)
-
数量:
5000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS99-04-7
Appearance (Color)White to pale yellow
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Melting Point (clear melt)108.0-115.0°C
FormCrystals or powder or crystalline powder or flakes
さらに表示
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluic acid is used for the production of N,N-di methyl formamide, toluene formyl chloride and toluene nitrile etc,.

Solubility
Soluble in alcohol and ether.

Notes
Incompatible with strong oxidizing agents. Store away from incompatible materials such as oxidizing agents.
RUO – Research Use Only

General References:

  1. Yong Ge; Frederic H Vaillancourt; Nathalie Y R Agar, Lindsay D Eltis. Reactivity of toluate dioxygenase with substituted benzoates and dioxygen. Journal of Bacteriology.2002, 184 (15), 4096-4103
  2. I Sarand, S Timonen, T Koivula, R Peltola, K Haahtela, R Sen, M Romantschuk. Tolerance and biodegradation of m-toluate by Scots pine, a mycorrhizal fungus and fluorescent pseudomonads individually and under associative conditions.Journal of Applied Microbiology.1999, 86 (5), 817-826