Ethyl 3,4,5-trimethylpyrrole-2-carboxylate, 98%
Ethyl 3,4,5-trimethylpyrrole-2-carboxylate, 98%
Ethyl 3,4,5-trimethylpyrrole-2-carboxylate, 98%
Thermo Scientific Chemicals

Ethyl 3,4,5-trimethylpyrrole-2-carboxylate, 98%

CAS: 2199-46-4 | C10H15NO2 | 181.235 g/mol
製品番号(カタログ番号) A13814.03
または、製品番号A13814-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2199-46-4
IUPAC Nameethyl 3,4,5-trimethyl-1H-pyrrole-2-carboxylate
Molecular FormulaC10H15NO2
InChI KeyWBOGFZDTCIQHSX-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(C)C(C)=C(C)N1
さらに表示
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)123.0-130.0?C
Appearance (Color)Pale pink
Octamethylporphyrin was readily obtained from ethyl 3,4,5-trimethylpyrrole-2-carboxylate. Steric control can be overridden by apparent electronic effect is demonstrated by the even bulkier pyrrole derivative, ethyl 3,4,5-trimethylpyrrole-2-carboxylate. The reaction with diazonium salts of theEthyl 3,4,5-trimethylpyrrole-2-carboxylatehas been examined in detail by chromatographic and spectroscopic methods. In the presence of a strong mineral acid, Ethyl 3,4,5-trimethylpyrrole-2-carboxylate reacts with p-nitrobenzenediazonium ions to give a red, chemically labile, azo dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octamethylporphyrin was readily obtained from ethyl 3,4,5-trimethylpyrrole-2-carboxylate. Steric control can be overridden by apparent electronic effect is demonstrated by the even bulkier pyrrole derivative, ethyl 3,4,5-trimethylpyrrole-2-carboxylate. The reaction with diazonium salts of theEthyl 3,4,5-trimethylpyrrole-2-carboxylatehas been examined in detail by chromatographic and spectroscopic methods. In the presence of a strong mineral acid, Ethyl 3,4,5-trimethylpyrrole-2-carboxylate reacts with p-nitrobenzenediazonium ions to give a red, chemically labile, azo dye.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Anita L. Arduini,; Norman M. Edelstein,; Jackie D. Jamerson,; John G. Reynolds,; Klaus. Schmid,; Josef. Takats. Syntheses and characterization of (.eta.5-C5H5)2U(NR2)2 compounds. Inorg. Chem., 1981, 20 (8), 2470-2474.
  2. S. Ramalingama,; S. Periandyb,; B. Narayananb,; S. Mohanc. FTIR and FTRaman spectroscopic investigation of 2-bromo-4-methylaniline using ab initio HF and DFT calculations. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2010, 76 (1), 84-92.