2-Hydroxyphenylacetic acid, 98%
2-Hydroxyphenylacetic acid, 98%
2-Hydroxyphenylacetic acid, 98%
2-Hydroxyphenylacetic acid, 98%
Thermo Scientific Chemicals

2-Hydroxyphenylacetic acid, 98%

CAS: 614-75-5 | C8H8O3 | 152.149 g/mol
製品番号(カタログ番号) A13924.18
または、製品番号A13924-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS614-75-5
IUPAC Name2-(2-hydroxyphenyl)acetic acid
Molecular FormulaC8H8O3
InChI KeyCCVYRRGZDBSHFU-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1O
さらに表示
Assay (HPLC)≥97.5% (U.K. Specification)
Assay from Supplier's CofA≥97.5%, unspecified (U.S. Specification)
CommentPurchased in U.K. and U.S.
Melting Point (clear melt)141.0-147.0°C (U.K. Specification)
Appearance (Color)Cream to yellow to pale brown
さらに表示
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

Solubility
Soluble in Methanol and Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. N Oureddine.; A Llouche,; S Ami S Ayadi. Synthesis of Hydroxytyrosol, 2-Hydroxyphenylacetic Acid, and 3-Hydroxyphenylacetic Acid by Differential Conversion of Tyrosol Isomers Using Serratia marcescens Strain. Journal of Agricultural and Food Chemistry. 2005, 53 6525-6430
  2. H. Kindl,; G. Billek. 2-Hydroxyphenylacetic Acid, a New Constituent of Higher Plant . Nature. 1962 194 579.