Cyclopropanemethylamine, 97%
Cyclopropanemethylamine, 97%
Cyclopropanemethylamine, 97%
Cyclopropanemethylamine, 97%
Thermo Scientific Chemicals

Cyclopropanemethylamine, 97%

CAS: 2516-47-4 | C4H9N | 71.123 g/mol
製品番号(カタログ番号) A13946.06
または、製品番号A13946-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2516-47-4
IUPAC Namecyclopropylmethanaminium
Molecular FormulaC4H10N
InChI KeyIGSKHXTUVXSOMB-UHFFFAOYSA-O
SMILES[NH3+]CC1CC1
さらに表示
Refractive Index1.4300-1.4350 @ 20°C (non-U.S. specification)
FormLiquid
Appearance (Color)Clear colorless
CommentPurchased in the U.S. and in other countries
Assay (GC)≥96.0% (non-U.S. specification)
さらに表示
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
RUO – Research Use Only
James R. Durig; Chao Zheng; Robin D. Warren; Peter Groner; Charles J. Wurrey; and Todor K. Gounev. Conformational and Structural Studies of Aminomethyl Cyclopropane from Temperature Dependent FT-IR Spectra of Rare Gas Solutions and ab Initio Calculations. J. Phys. Chem. A, 2003, 107 (39), 7713-7726.