Thermo Scientific™

4-Fluoro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A13977.14
25 g, Each
Thermo Scientific™

4-Fluoro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

Catalog number: A13977.14
25 g, Each
Quantity
Catalog number: A13977.14
also known as A13977-14
Price (USD)
Price: 305.00
Online price: 263.65
Your price:
Quantity
-

Chemical Identifiers

CAS
367-31-7
IUPAC Name
4-fluorobenzene-1,2-diamine
Molecular Formula
C6H7FN2
InChI Key
KWEWNOOZQVJONF-UHFFFAOYSA-N
SMILES
NC1=CC=C(F)C=C1N
Appearance (Color)
Cream to brown
Form
Powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
89.0-95.0?C

Description

4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-o-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text