Hydrazine monohydrate, 98+%
Hydrazine monohydrate, 98+%
Hydrazine monohydrate, 98+%
Thermo Scientific Chemicals

Hydrazine monohydrate, 98+%

CAS: 7803-57-8 | H6N2O | 50.06 g/mol
数量:
50 g
250 g
2500 g
製品番号(カタログ番号) A14005.30
または、製品番号A14005-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7803-57-8
Appearance (Color)Clear colorless
FormLiquid
Assay (Hydrazine)≥98.0 to ≤102.0% (non-U.S. specification)
Assay from Suppliers CofA≥98.0% (U.S. specifiactionl)
CommentSpecification differs for U.S. and non-U.S. material where indicated
さらに表示
Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrazine monohydrate is used as a reducing agent. It is also used in water treatment, especially effluents and industrial boilers, metal treatment and mine extraction process, and synthesis of active ingredients in the pharmaceutical industry like 3-amino-1,2,4-triazole cefazolin, rizatriptan, anastrozole, fluconazole, metazachlor metamitron pyrazole, metribuzin, paclobutrazol, diclobutrazole, propiconazole, and triadimefon. It is involved in the preparation of heterocycles like pyrazoles and pyridazines. It is actively involved in the Wolff-Kishner reduction, which converts the carbonyl group of a ketone into a methylene bridge through a hydrazone intermediate. Further, it is used to cleave N-alkylated phthalimide derivatives. In the Einhorn-Brunner reaction, it reacts with imides to get triazoles.

Solubility
Miscible with water and alcohol. Immiscible with chloroform, hydrocarbons and ether.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.comIncompatible with oxidizing agents, oxygen, copper, organic materials and zinc.
RUO – Research Use Only

General References:

  1. Forms stable, water-free solid complexes consisting of one hydrazine molecule associated with one molecule of hydroquinone or two molecules of 4-methoxyphenol. These have potential as replacements for the very hazardous anhydrous hydrazine, e.g. in the solid state reaction with esters to give pure hydrazides: J. Chem. Soc., Chem. Commun., 1531, (1995).
  2. Diimide precursor: for generation of diimide by oxidation with O2, and use in selective reduction of a double bond in the presence of a cyclopropane ring, see: J. Chem. Soc., Chem. Commun., 525 (1973). Review of diimide reductions: Org. React., 40, 91 (1991). See also Hydroxyl amine-O-sulfonic acid, A12560 .
  3. In the Wolff-Kishner carbonyl to methylene reduction (heating with hydrazine and alkali hydroxide), diethylene glycol is now the usual solvent (the Huang Minlon modification): J. Am. Chem. Soc., 68, 2487 (1946); Org. Synth. Coll., 4, 510 (1963). The reduction has also been performed at ambient temperature by the use of KO-t-Bu in DMSO: J. Am. Chem. Soc., 84, 1734 (1962). See also Isatin, A12468 for an analogous reduction.
  4. In the presence of Rh on carbon, nitrobenzene is selectively reduced to phenylhydroxylamine: Org. Synth. Coll., 8, 16 (1993).
  5. For a brief feature on uses of the reagent in synthesis, see: Synlett, 2445 (2004).
  6. Shiraishi, Y.; Hirakawa, H.; Hirai, T. Photocatalytic Hydrogenation of Nitroaromatics to Anilines on Silica-Supported Iron Oxides with Hydrazine Monohydrate as a Reductant. J. Chem. Eng. Jpn. 2015, 48 (2), 141-146.
  7. Sun, J. K.; Xu, Q. Metal Nanoparticles Immobilized on Carbon Nanodots as Highly Active Catalysts for Hydrogen Generation from Hydrazine in Aqueous Solution. Chem. Cat. Chem. 2015, 7 (3), 526-531.