Thermo Scientific Chemicals

1-Bromo-3-chlorobenzene, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14026.36
500 g, Each
Thermo Scientific Chemicals

1-Bromo-3-chlorobenzene, 99%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14026.36
500 g, Each
数量
製品番号(カタログ番号): A14026.36
または、製品番号A14026-36

化学物質識別子

CAS
108-37-2
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.5750-1.5790 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

概要

1-Bromo-3-chlorobenzene undergoes bromine-magnesium exchange reactions with i-PrMgCl-LiCl in THF at 0°C and is strongly accelerated by electron-withdrawing substituents. It acts as test substrate to study the application of a new catalytic system for cyanation of various aryl halides using K4[Fe(CN)6] as cyanating source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-chlorobenzene undergoes bromine-magnesium exchange reactions with i-PrMgCl-LiCl in THF at 0°C and is strongly accelerated by electron-withdrawing substituents. It acts as test substrate to study the application of a new catalytic system for cyanation of various aryl halides using K4[Fe(CN)6] as cyanating source.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text