2,4-Difluorophenol, 98+%
2,4-Difluorophenol, 98+%
2,4-Difluorophenol, 98+%
2,4-Difluorophenol, 98+%
Thermo Scientific Chemicals

2,4-Difluorophenol, 98+%

CAS: 367-27-1 | C6H4F2O | 130.09 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A14044.18
または、製品番号A14044-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS367-27-1
IUPAC Name2,4-difluorophenol
Molecular FormulaC6H4F2O
InChI KeyNVWVWEWVLBKPSM-UHFFFAOYSA-N
SMILESOC1=CC=C(F)C=C1F
さらに表示
FormFused solid or clear liquid as melt
Refractive Index1.4835-1.4880 @ 20°C (non-U.S. specification)
Appearance (Color)Colorless to white to pale yellow
Assay (GC)≥98.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only