Thermo Scientific Chemicals

2-Amino-4-methylpyridine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14177.22
100 g, Each
Thermo Scientific Chemicals

2-Amino-4-methylpyridine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号): A14177.22
100 g, Each
数量
製品番号(カタログ番号): A14177.22
または、製品番号A14177-22

化学物質識別子

CAS
695-34-1
IUPAC Name
4-methylpyridin-2-amine
Molecular Formula
C6H8N2
InChI Key
ORLGLBZRQYOWNA-UHFFFAOYSA-N
SMILES
CC1=CC=NC(N)=C1
Form
Crystals or powder or crystalline powder or crystalline flakes
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
<1.0%
Appearance (Color)
White to pale yellow or pale cream to cream or pale pink or pale brown
Identification (FTIR)
Conforms

概要

2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methyl­pyridinium 2-hy­droxy­benzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.

Solubility
Soluble in water (41 mg/ml at 20°C), lower alcohols (slightly), chloroform, ethyl acetate, and DMF.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water. Store at -20°C.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text