2-Fluorobenzenesulfonyl chloride, 98%
2-Fluorobenzenesulfonyl chloride, 98%
2-Fluorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2-Fluorobenzenesulfonyl chloride, 98%

CAS: 2905-21-7 | C6H4ClFO2S | 194.60 g/mol
製品番号(カタログ番号) A14181.03
または、製品番号A14181-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2905-21-7
IUPAC Name2-fluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H4ClFO2S
InChI KeyZSZKAQCISWFDCQ-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1S(Cl)(=O)=O
さらに表示
Refractive Index1.5350-1.5390 @ 20?C
FormLiquid as melt
Appearance (Color)Clear, colorless to yellow
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only