Thermo Scientific Chemicals

L-Glutamine, 99%, Thermo Scientific Chemicals

Catalog number: A14201.30
250 g, Each
Thermo Scientific Chemicals

L-Glutamine, 99%, Thermo Scientific Chemicals

Catalog number: A14201.30
250 g, Each
Quantity
Catalog number: A14201.30
also known as A14201-30
Price (USD)
Price: 119.00
Online price: 101.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56-85-9
IUPAC Name
(2S)-2-amino-4-carbamoylbutanoic acid
Molecular Formula
C5H10N2O3
InChI Key
ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES
N[C@@H](CCC(N)=O)C(O)=O
Optical Rotation
33.0 ± 1.0? (C=5 in 5N HCl)
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Form
Powder
Identification (FTIR)
Conforms

Description

L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in methanol, ether, benzene, acetone, ethyl acetate and chloroform.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text