Thermo Scientific Chemicals

Bis(2-chloroethyl) ether, 99%, Thermo Scientific Chemicals

Catalog number: A14234.30
250 g, Each
Thermo Scientific Chemicals

Bis(2-chloroethyl) ether, 99%, Thermo Scientific Chemicals

Catalog number: A14234.30
250 g, Each
Quantity
Catalog number: A14234.30
also known as A14234-30
Price (USD)
Price: 79.00
Online price: 68.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-44-4
IUPAC Name
1-chloro-2-(2-chloroethoxy)ethane
Molecular Formula
C4H8Cl2O
InChI Key
ZNSMNVMLTJELDZ-UHFFFAOYSA-N
SMILES
ClCCOCCCl
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4555-1.4585 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl) ether is used as a solvent and soil fumigant. It may be used in the preparation of macrocyclic polyethers, dibenzo-18-crown-6 polyether and dicyclohexyl-18-crown-6 polyether.

Solubility
Soluble in water. (1.1 g/L) at 20°C.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text