Thermo Scientific Chemicals

3-Acetylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14246.22
100 g, Each
Thermo Scientific Chemicals

3-Acetylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: A14246.22
100 g, Each
Quantity
Catalog number: A14246.22
also known as A14246-22
Price (USD)
Price: 118.00
Online price: 100.65
Your price:
Quantity
-

Chemical Identifiers

CAS
350-03-8
IUPAC Name
1-(pyridin-3-yl)ethan-1-one
Molecular Formula
C7H7NO
InChI Key
WEGYGNROSJDEIW-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=CN=C1
Identification (FTIR)
Conforms
Refractive Index
1.5330-1.5370 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥97.5%

Description

3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Acetylpyridine used as an intermediate for the synthesis of risedronate sodium, inhibitor of bone resorption. It is also used in perfumery.

Solubility
Soluble in Chloroform. Miscible with water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text