Bathophenanthroline, 98+%
Bathophenanthroline, 98+%
Bathophenanthroline, 98+%
Thermo Scientific Chemicals

Bathophenanthroline, 98+%

CAS: 1662-01-7 | C24H16N2 | 332.406 g/mol
製品番号(カタログ番号) A14258.03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1662-01-7
IUPAC Name4,7-diphenyl-1,10-phenanthroline
Molecular FormulaC24H16N2
InChI KeyDHDHJYNTEFLIHY-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)Pale cream to cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)217.0-224.0?C
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for iron.
  2. Ramos, P. M. R.; Silva, P. S. P.; Posada, P. C.; Silva, M. R.; Milne, B. F.; Nogueira, F.; Gil, J. M. Synthesis, structure, theoretical studies and luminescent properties of a ternary erbium(III) complex with acetylacetone and bathophenanthroline ligands. J. Lumin. 2015, 162, 41-49.
  3. Mazumdar, P.; Das, D.; Sahoo, G. P.; Moran, G. S.; Misra, A. Aggregation induced emission enhancement from Bathophenanthroline microstructures and its potential use as sensor of mercury ions in water. Phys. Chem. Chem. Phys. 2014, 16 (13), 6283-6293.