Thermo Scientific Chemicals

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

產品號碼: A14268.09
10 g, Each
Thermo Scientific Chemicals

Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals

產品號碼: A14268.09
10 g, Each
Quantity
產品號碼: A14268.09
價格 (TWD)

化學識別

CAS
756-79-6
Appearance (Color)
Clear colorless
Assay (GC)
≥ 96.0%
Form
Liquid

說明

Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl methylphosphonate is used as a catalyst and a reagent in organic synthesis for the conversion of esters to ketophosphonates. It is an additive for the synthesis of unsaturated polyester resin which has flame retardant high phosphorous and UV-cured epoxy acrylate. It finds application as hydraulic fluids as well.

Solubility
Miscible with water, ethanol, ether, alcohol, benzene, acetone and carbon tetrachloride. Immiscible with heavy mineral oil.

Notes
Incompatible materials strong oxidizing agents and strong bases.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text