Magnesium hydrogen phosphate trihydrate, 97+%
Magnesium hydrogen phosphate trihydrate, 97+%
Magnesium hydrogen phosphate trihydrate, 97+%
Magnesium hydrogen phosphate trihydrate, 97+%
Thermo Scientific Chemicals

Magnesium hydrogen phosphate trihydrate, 97+%

CAS: 7782-75-4 | H7MgO7P | 174.33 g/mol
製品番号(カタログ番号) A14279.30
または、製品番号A14279-30
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7782-75-4
IUPAC Namemagnesium(2+) trihydrate hydrogen phosphate
Molecular FormulaH7MgO7P
InChI KeyOKIWLDVQGKRUNR-UHFFFAOYSA-L
SMILESO.O.O.[Mg++].OP([O-])([O-])=O
さらに表示
Complexometric Titration≥97.0 to ≤103.0%
Identification (FTIR)Conforms
Loss on ignition29.0-36.5%
FormPowder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only