1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
1,2,4,5-Tetrachlorobenzene, 98%
Thermo Scientific Chemicals

1,2,4,5-Tetrachlorobenzene, 98%

CAS: 95-94-3 | C6H2Cl4 | 215.882 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A14318.18
または、製品番号A14318-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-94-3
IUPAC Name1,2,4,5-tetrachlorobenzene
Molecular FormulaC6H2Cl4
InChI KeyJHBKHLUZVFWLAG-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(Cl)C=C1Cl
さらに表示
Melting Point (clear melt)137.5-143.5?C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)White to pale pink to grey
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

Solubility
Insoluble in water. Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Peter Sander, et.al. Degradation of 1,2,4-Trichloro- and 1,2,4,5-Tetrachlorobenzene by Pseudomonas Strains.Appl. Environ. Microbiol.,1991,57(5), 1430-1440.
  2. Stefan Beil, et al. Genetic and Biochemical Characterization of the Broad Spectrum Chlorobenzene Dioxygenase from Burkholderia Sp. Strain PS12 — Dechlorination of 1,2,4,5-Tetrachlorobenzene.Eur. J. Biochem.,1997,247(1), 190-199.