N-Phenylanthranilic acid, 99%
N-Phenylanthranilic acid, 99%
N-Phenylanthranilic acid, 99%
Thermo Scientific Chemicals

N-Phenylanthranilic acid, 99%

CAS: 91-40-7 | C13H11NO2 | 213.236 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A14340.14
または、製品番号A14340-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS91-40-7
IUPAC Name2-(phenylamino)benzoic acid
Molecular FormulaC13H11NO2
InChI KeyZWJINEZUASEZBH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1NC1=CC=CC=C1
さらに表示
FormPowder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream to yellow to green to grey
さらに表示
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DPC is a potent, non-specific blocker of Cl- channel and chloride-bicarbonate exchange. DPC also inhibits CFTR (cystic fibrosis transmembrane regulator) ATPase activity. It is used to detect vanadium in steel.

Solubility
Soluble in ethanol, DMSO (50 mg/ml), hot alcohol, and methanol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Maite L. Docampo Palaciosa and Rolando F. Pellón Comdom. Synthesis of N-Phenylanthranilic Acid Derivatives Using Water as Solvent in the Presence of Ultrasound Irradiation.Synth. Commun.,2003,33(10), 1771-1775.
  2. Ali Almasirad, et al. Synthesis and Analgesic Activity of 2-Phenoxybenzoic Acid and N-Phenylanthranilic Acid Hydrazides.Biol. Pharm. Bull.,2006,29(6), 1180-1185.