3,5-Dimethoxybenzoic acid, 99%
3,5-Dimethoxybenzoic acid, 99%
3,5-Dimethoxybenzoic acid, 99%
3,5-Dimethoxybenzoic acid, 99%
Thermo Scientific Chemicals

3,5-Dimethoxybenzoic acid, 99%

CAS: 1132-21-4 | C9H10O4 | 182.18 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A14374.36
または、製品番号A14374-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1132-21-4
IUPAC Name3,5-dimethoxybenzoic acid
Molecular FormulaC9H10O4
InChI KeyIWPZKOJSYQZABD-UHFFFAOYSA-N
SMILESCOC1=CC(=CC(OC)=C1)C(O)=O
さらに表示
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Melting Point (clear melt)181.0-187.0°C
FormPowder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only