4-Methoxyphenylacetic acid, 98+%
4-Methoxyphenylacetic acid, 98+%
4-Methoxyphenylacetic acid, 98+%
4-Methoxyphenylacetic acid, 98+%
Thermo Scientific Chemicals

4-Methoxyphenylacetic acid, 98+%

CAS: 104-01-8 | C9H10O3 | 166.176 g/mol
数量:
5 g
100 g
500 g
製品番号(カタログ番号) A14399.36
または、製品番号A14399-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS104-01-8
IUPAC Name2-(4-methoxyphenyl)acetic acid
Molecular FormulaC9H10O3
InChI KeyNRPFNQUDKRYCNX-UHFFFAOYSA-N
SMILESCOC1=CC=C(CC(O)=O)C=C1
さらに表示
Identification (FTIR)Conforms
Appearance (Color)White to cream or pale yellow
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystalline powder and/or chunks
Melting Point (clear melt)84-88?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only