Thermo Scientific Chemicals

Aniline, 99+%, Thermo Scientific Chemicals

Catalog number: A14443.0I
5000 g, Each
Thermo Scientific Chemicals

Aniline, 99+%, Thermo Scientific Chemicals

Catalog number: A14443.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A14443.0I
also known as A14443-0I
Price (USD)
Price: 212.00
Online price: 180.65
Your price:
Quantity
-

Chemical Identifiers

CAS
62-53-3
IUPAC Name
aniline
Molecular Formula
C6H7N
InChI Key
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1
Refractive Index
1.5845-1.5875 @ 20?C
Appearance (Color)
Clear, colorless to yellow or pale orange to pale brown
Form
Liquid
Assay (GC)
≥99.0%
Comment
May darken on storage

Description

Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents, iron and iron salts.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text