Iodoethane, 98+%, stab. with copper
Iodoethane, 98+%, stab. with copper
Iodoethane, 98+%, stab. with copper
Iodoethane, 98+%, stab. with copper
Thermo Scientific Chemicals

Iodoethane, 98+%, stab. with copper

CAS: 75-03-6 | C2H5I | 155.966 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A14444.22
または、製品番号A14444-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-03-6
IUPAC Nameiodoethane
Molecular FormulaC2H5I
InChI KeyHVTICUPFWKNHNG-UHFFFAOYSA-N
SMILESCCI
さらに表示
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5100-1.5140 @ 20?C
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoethane is an excellent ethylating agent used in organic synthesis to introduce an ethyl group into a compound. It reacts with magnesium to form the Grignard reagent, ethylmagnesium iodide which is used in organic synthesis. It is involved in the preparation of 1-ethyl-3-nitro-2-phenyl-indole and also serves as hydrogen radical promoter.

Solubility
Miscible with water.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and magnesium.
RUO – Research Use Only

General References:

  1. Nijamudheen, A.; Datta, A. Mechanism for C-I Bond Dissociation in Iodoethane, Iodobenzene, and Iodoethene for the C-C Cross Coupling Reactions over Gold Clusters. J. Phys. Chem. C 2013, 117 (41), 21433-21440.
  2. Ljubic, I.; Matasovic, B.; Bonifacic, M. An efficient buffer-mediated control between free radical substitution and proton-coupled electron transfer: dehalogenation of iodoethane by the alpha-hydroxyethyl radical in aqueous solution. Phys. Chem. Chem. Phys. 2013, 15 (41), 18001-18011.