2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D
Thermo Scientific Chemicals

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D

CAS: 24493-21-8 | C6H13NaO2Si | 172.267 g/mol
数量:
250 mg
1 g
5 g
製品番号(カタログ番号) A14489.03
または、製品番号A14489-03
価格(JPY)
-
見積もりを依頼する
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS24493-21-8
IUPAC Namesodium 3-(trimethylsilyl)(²H₄)propanoate
Molecular FormulaC6H13NaO2Si
InChI KeyOIIWPAYIXDCDNL-HGFPCDIYSA-M
SMILES[Na+].[2H]C([2H])(C([O-])=O)C([2H])([2H])[Si](C)(C)C
さらに表示
Elemental Analysis≥98.0 atom% deuterium
Water Content (Karl Fischer Titration)≤2.0%
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥92.5% to ≤107.5%
Identification (FTIR)Conforms
さらに表示
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Priyadarshi Ranjan; Ashish Gupta; Sandeep Kumar; Gowda Ajjapa Nagan Gowda; Aseem Ranjan; Abhinav Arun Sonker; Abhijit Chandra; Rama Kant. Detection of new amino acid markers of liver trauma by proton nuclear magnetic resonance spectroscopy. Liver International. 2006, 26, (6),703-707
  2. Ashish Gupta; Mayank Dwivedi; G.A.Nagana Gowda, Archana Ayyagari; A.A. Mahdi; M.Bhandari; C.L.Khetrapal. 1H NMR spectroscopy in the diagnosis of Pseudomonas aeruginosa-induced urinary tract infection. NMR in Biomedicine. 2005, 18, (5),293-299
  3. Water-soluble NMR reference standard.