This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Thymol is used as a preservative in halothane. It acts as an anesthetic, antiseptic in mouth wash, stabilizer in pharmaceutical preparations. It inhibits the growth and lactate production as well as reduces the uptake of cellular glucose within the bacteria. It is an active ingredient in toothpastes like euthymol. It is involved to control varroa mites and prevent fermentation and the growth of mold in bee colonies.
Solubility
Slightly soluble in water. Soluble in chloroform, alcohol and ether.
Notes
Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only
General References:
- Mohan, R. S.; Onsen, L. T. Isolation of Thymol from Carom Seeds. Chem. Educ. 2015, 20, 72-73.
- Aznar, A.; Fernández, P. S.; Periago, P. M.; Palop, A. Antimicrobial activity of nisin, thymol, carvacrol and cymene against growth of Candida lusitaniae. Food Sci. Technol. Int. 2015, 21 (1), 72-79.