Thermo Scientific Chemicals

2-Iodoethanol, 99%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A14658.22
100 g, Each
Thermo Scientific Chemicals

2-Iodoethanol, 99%, stab. with copper, Thermo Scientific Chemicals

Catalog number: A14658.22
100 g, Each
Quantity
Catalog number: A14658.22
also known as A14658-22
Price (USD)
Price: 403.00
Online price: 342.65
Your price:
Quantity
-

Chemical Identifiers

CAS
624-76-0
IUPAC Name
2-iodoethan-1-ol
Molecular Formula
C2H5IO
InChI Key
QSECPQCFCWVBKM-UHFFFAOYSA-N
SMILES
OCCI
Form
Liquid
Refractive Index
1.5695-1.5735 @ 20?C
Appearance (Color)
Clear colorless to yellow
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text