6-Benzyladenine, 99%
6-Benzyladenine, 99%
6-Benzyladenine, 99%
Thermo Scientific Chemicals

6-Benzyladenine, 99%

CAS: 1214-39-7 | C12H11N5 | 225.26 g/mol
製品番号(カタログ番号) A14678.03
または、製品番号A14678-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1214-39-7
IUPAC NameN-benzyl-7H-purin-6-amine
Molecular FormulaC12H11N5
InChI KeyNWBJYWHLCVSVIJ-UHFFFAOYSA-N
SMILESC(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
さらに表示
Melting Point (clear melt)227.5-234.5°C
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.

Notes
Incompatible with strong oxidizing agent.
RUO – Research Use Only

General References:

  1. Bound, S. A. Optimising crop load and fruit quality of ‘Packham’s Triumph’ pear with ammonium thiosulfate, ethephon and 6-benzyladenine. Sci. Hortic. 2015, 192, 187-196.
  2. Kaviani, B.; Hashemabadi, D.; Khodabakhsh, H.; Onsinejad, R.; Ansari, M. H.; Haghighat, N. Micropropagation of Begonia rex Putz. by 6-benzyladenine and alpha-naphthalene acetic acid. Int. J. Biosci. 2015, 6 (5), 8-15.