Cyclohexanecarboxylic acid, 98%
Cyclohexanecarboxylic acid, 98%
Cyclohexanecarboxylic acid, 98%
Cyclohexanecarboxylic acid, 98%
Thermo Scientific Chemicals

Cyclohexanecarboxylic acid, 98%

CAS: 98-89-5 | C7H12O2 | 128.171 g/mol
製品番号(カタログ番号) A14693.30
または、製品番号A14693-30
価格(JPY)
-
見積もりを依頼する
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS98-89-5
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
FormCrystals or powder or crystalline powder or fused solid or chunks
Appearance (Color)White
Melting Point (clear melt)27-35?C
Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarboxylic acid can be used to determine complex binding constants of the three native cyclodextrins with seven cyclohexane derivatives.

Solubility
Soluble in water 0.201 g/100 mL (15°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. RJ Williams.; WC Evans. The metabolism of benzoate by Moraxella species through anaerobic nitrate respiration. Evidence for a reductive pathway. Biochem. j. 19757 (1), 43-49.
  2. BM Babior.; K Bloch. Aromatization of cyclohexanecarboxylic acid. Journal of Biological Chemistry. 1966241 (1), 3643-3651.