Thermo Scientific Chemicals

4-Chlorobenzyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A14696.0B
1000 g, Each
Thermo Scientific Chemicals

4-Chlorobenzyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A14696.0B
1000 g, Each
Quantity
Catalog number: A14696.0B
also known as A14696-0B
Price (USD)
Price: 190.00
Online price: 161.65
Your price:
Quantity
-

Chemical Identifiers

CAS
104-83-6
IUPAC Name
1-chloro-4-(chloromethyl)benzene
Molecular Formula
C7H6Cl2
InChI Key
JQZAEUFPPSRDOP-UHFFFAOYSA-N
SMILES
ClCC1=CC=C(Cl)C=C1
Appearance (Color)
White or clear colorless to pale yellow
Refractive Index
1.5555-1.5595 @ 20?C
Form
Fused solid or liquid as melt
Assay (GC)
≥98.0%

Description

4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzyl chloride was the test compound for determining rat hepatocellular thiols by HPLC. As a reagent used in derivatization of, for example, a calix[4]arene and a tryptamine. It is widely used as an intermediate for manufacturing organic compounds like benzyl alcohol and benzyl cyanide. Also used in fuel as a gum inhibitor.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text