trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals
trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals
trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Methyl-beta-nitrostyrene, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
A14721.06
または、製品番号A14721-06
5 g
製品番号(カタログ番号) A14721.06
または、製品番号A14721-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5153-68-4
IUPAC Name1-methyl-4-(2-nitroethenyl)benzene
Molecular FormulaC9H9NO2
InChI KeyJSPNBERPFLONRX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C[N+]([O-])=O)C=C1
さらに表示
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)100.0-106.0?C
trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Methyl-β-nitrostyrene may be used as a reagent in the synthesis of N-benzylpyrrolomorphinans. It is also used as an organic intermediate for pharmaceutical.

Solubility
Sparingly Soluble in water (0.070 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. JP Wan.; Y Liu. Recent advances in new multicomponent synthesis of structurally diversified 1, 4-dihydropyridines. RSC Advances. 20122 (1), 9763-9777.
  2. RW Layer. The Chemistry of Imines. Chemical Reviews, . 196363 (5), 489-510.