4-Bromo-o-xylene, 97%
4-Bromo-o-xylene, 97%
4-Bromo-o-xylene, 97%
Thermo Scientific Chemicals

4-Bromo-o-xylene, 97%

CAS: 583-71-1 | C8H9Br | 185.064 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A14736.22
または、製品番号A14736-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS583-71-1
IUPAC Name4-bromo-1,2-dimethylbenzene
Molecular FormulaC8H9Br
InChI KeyQOGHRLGTXVMRLM-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)C=C1C
さらに表示
Appearance (Color)Clear colorless
Refractive Index1.5535-1.5585 @ 20?C
Assay (GC)≥96.0%
FormLiquid
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. M.Arivazhagan; R.Meenakshi. Vibrational spectroscopic studies and DFT calculations of 4-bromo-o-xylene. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2012, 91, 419-430
  2. G.A.Bagirzade. Kinetics of oxidative ammonolysis of 4-bromo-o-xylene: I. Transformations of 4-bromo-o-xylene and 4-bromo-o-tolunitrile. Russian Journal of General Chemistry. 2010, 80, (8),1672-1676