3-Chlorophenylhydrazine hydrochloride, 97%
3-Chlorophenylhydrazine hydrochloride, 97%
3-Chlorophenylhydrazine hydrochloride, 97%
3-Chlorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

3-Chlorophenylhydrazine hydrochloride, 97%

CAS: 2312-23-4 | C6H8Cl2N2 | 179.044 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A14770.09
または、製品番号A14770-09
価格(JPY)
-
見積もりを依頼する
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2312-23-4
IUPAC Namehydrogen (3-chlorophenyl)hydrazine chloride
Molecular FormulaC6H8Cl2N2
InChI KeyCRRIAWUJYMLJOE-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=CC(Cl)=C1
さらに表示
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only