Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl methylphosphonate, 97%, Thermo Scientific Chemicals

数量:
10 g
25 g
50 g
製品番号(カタログ番号) A14772.18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS683-08-9
IUPAC Namediethyl methylphosphonate
Molecular FormulaC5H13O3P
InChI KeyNYYLZXREFNYPKB-UHFFFAOYSA-N
SMILESCCOP(C)(=O)OCC
さらに表示
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4115-1.4165 @ 20?C
Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl methylphosphonate is used as a precursor for the synthesis of gem-difluroalkenyphosphonate via reaction with alpha-trifluromethylstyrene. It is also used in the synthesis of phosphonylated peptides, fluoroalkyl alpha- and beta-aminophosphonates, pyridone alkaloids with neuritogenic activity and lipophilic meropenem-derived prodrugs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. For synthetic applications of the lithio-derivatives of esters of methanephosphonic acid, see Dimethyl methyl phosphonate, A14268.
  2. Dialkylation has been used in the synthesis of cyclobutanephosphonates which are intermediates in the synthesis of 3-phosphonocyclobutyl amino acids: J. Chem. Soc., Perkin 1, 493 (1997).
  3. Reaction of the lithio-derivative with dialkyl cyanamides gives, instead of the expected 2-phosphorylacetamide, a rearranged N-phosphorylated amidine: Chem. Commun., 609 (1998):
  4. Mattea, F.; Peters, C. J.; Kroon, M. C. Experimental Determination of the Solubilities of CO2 and CH4 in Diethyl Methylphosphonate. J. Chem. Eng. Data 2011, 56 (6), 2960-2963.
  5. JeongaJang, Y.; Murale, D. P.; Churchill, D. G. Novel reversible and selective nerve agent simulant detection in conjunction with superoxide turn-on probing. Analyst 2014, 139 (7), 1614-1617.