5-Bromouracil, 98+%
5-Bromouracil, 98+%
5-Bromouracil, 98+%
Thermo Scientific Chemicals

5-Bromouracil, 98+%

CAS: 51-20-7 | C4H3BrN2O2 | 190.98 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A14799.06
または、製品番号A14799-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS51-20-7
IUPAC Name5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H3BrN2O2
InChI KeyLQLQRFGHAALLLE-UHFFFAOYSA-N
SMILESBrC1=CNC(=O)NC1=O
さらに表示
Appearance (Color)White to cream to pale brown
Assay (HPLC)≥98.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only