Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- Ludwig Bauer.; Thomas L. Welsh. The Synthesis of Derivatives of α-Mercaptoamidines1,2. J. Org. Chem. 1962, 27, (12), 4382-4385.
- Everett M. Stone.; Terezie H. Schaller.; Helena Bianchi.; Maria D. Person.; Walter Fast. Inactivation of Two Diverse Enzymes in the Amidinotransferase Superfamily by 2-Chloroacetamidine: Dimethylargininase and Peptidylarginine Deiminase. Biochemistry. 1971, 44, (12), 13744-13752.