Thioanisole, 99%
Thioanisole, 99%
Thioanisole, 99%
Thermo Scientific Chemicals

Thioanisole, 99%

CAS: 100-68-5 | C7H8S | 124.201 g/mol
数量:
25 g
100 g
500 g
2500 g
製品番号(カタログ番号) A14846.22
または、製品番号A14846-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100-68-5
IUPAC Name(methylsulfanyl)benzene
Molecular FormulaC7H8S
InChI KeyHNKJADCVZUBCPG-UHFFFAOYSA-N
SMILESCSC1=CC=CC=C1
さらに表示
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.5850-1.5880 @ 20?C
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Monolithiation is thought to occur initially on the ring, but rearrangement then gives the more stable phenylthiomethyllithium: J. Org. Chem., 31, 4097 (1966); Tetrahedron Lett., 1961 (1977). Further metallation occurs at the ortho-position. See, e.g.: Tetrahedron, 46, 861 (1990). Demetallation and acylation is the basis of a route to 3-substituted benzo[b]thiophenes: Synthesis, 888 (1988):
  2. Cation scavenger in the acidic deprotection of peptides; see, e.g.: Int. J. Pept. Prot. Res., 36, 255 (1990). See Appendix 6. The combination of the soft nucleophile thioanisole and the hard acid triflic acid has been used for the cleavage of methyl ethers, e.g. O-methyltyrosine, resistant to other methods: J. Chem. Soc., Chem. Commun., 971 (1979). The same conditions have been used for the cleavage of Cbz, Boc, benzyl and 4-methoxybenzyl protecting groups in peptide synthesis: Chem. Pharm. Bull., 28, 1214 (1980); 29, 600 (1981); J. Chem. Soc., Chem. Commun., 101 (1980); see also: Angew. Chem. Int. Ed., 33, 1729 (1994).
  3. Trehoux, A.; Roux, Y.; Guillot, R.; Mahy, J. P.; Avenier, F. Catalytic oxidation of dibenzothiophene and thioanisole by a diiron(III) complex and hydrogen peroxide. J. Mol. Catal. A: Chem. 2015, 396, 40-46.
  4. Li, S. L.; Xu, X.; Truhlar, D. G. Computational simulation and interpretation of the low-lying excited electronic states and electronic spectrum of thioanisole. Phys. Chem. Chem. Phys. 2015, 17 (31), 20093-20099.