4-Bromo-2,6-dichloroaniline, 98%
4-Bromo-2,6-dichloroaniline, 98%
4-Bromo-2,6-dichloroaniline, 98%
Thermo Scientific Chemicals

4-Bromo-2,6-dichloroaniline, 98%

CAS: 697-88-1 | C6H4BrCl2N | 240.909 g/mol
製品番号(カタログ番号) A14884.22
または、製品番号A14884-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS697-88-1
IUPAC Name4-bromo-2,6-dichloroaniline
Molecular FormulaC6H4BrCl2N
InChI KeyNPQBZKNXJZARBJ-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=C(Br)C=C1Cl
さらに表示
Melting Point (clear melt)83.0-89.0?C
Appearance (Color)Cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only